ADVH49B8340D AChemBlock

8-fluorochroman-4-amine

  • Cas Number : 791043-28-2
  • Molecular Weight : 167.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/8-fluorochroman-4-amine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S88002
Availability details for
Quantity
  • Availability
  • Add to Cart