ADVH49B83410 AChemBlock

1-(4-bromo-2-fluorophenyl)-2-methylpropan-1-one

  • Cas Number : 801303-32-2
  • Molecular Weight : 245.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/1-4-bromo-2-fluorophenyl-2-methylpropan-1-one.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S88005
Availability details for
Quantity
  • Availability
  • Add to Cart