ADVH49B83411 AChemBlock

6-fluoronaphthalen-1-ol

  • Cas Number : 804498-72-4
  • Molecular Weight : 162.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/6-fluoronaphthalen-1-ol.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S88006
Availability details for
Quantity
  • Availability
  • Add to Cart