ADVH49B83413 AChemBlock

(3-((pyridin-4-ylthio)methyl)phenyl)methanol

  • Cas Number : 811801-40-8
  • Molecular Weight : 231.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/3-pyridin-4-ylthio-methyl-phenyl-methanol.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S88008
Availability details for
Quantity
  • Availability
  • Add to Cart