ADVH49B83417 AChemBlock

4-methyl-2,3-dihydro-1H-indene

  • Cas Number : 824-22-6
  • Molecular Weight : 132.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/4-methyl-2-3-dihydro-1h-indene.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S88012
Availability details for
Quantity
  • Availability
  • Add to Cart