ADVH49B83418 AChemBlock

(R)-1-(naphthalen-2-yl)ethan-1-amine hydrochloride

  • Cas Number : 82572-03-0
  • Molecular Weight : 207.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/r-1-naphthalen-2-yl-ethan-1-amine-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S88013
Availability details for
Quantity
  • Availability
  • Add to Cart