ADVH49B8341A AChemBlock

8-chloro-3,4-dihydroquinolin-2(1H)-one

  • Cas Number : 83229-23-6
  • Molecular Weight : 181.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/8-chloro-3-4-dihydroquinolin-2-1h-one.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S88015
Availability details for
Quantity
  • Availability
  • Add to Cart