ADVH49B8341F AChemBlock

(R)-1-(3-bromophenyl)-2,2,2-trifluoroethan-1-amine hydrochloride

  • Cas Number : 842169-97-5
  • Molecular Weight : 290.51

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/r-1-3-bromophenyl-2-2-2-trifluoroethan-1-amine-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S88020
Availability details for
Quantity
  • Availability
  • Add to Cart