ADVH49B83420 AChemBlock

1-(4-bromophenyl)-2,2,2-trifluoroethan-1-amine

  • Cas Number : 843608-46-8
  • Molecular Weight : 254.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/1-4-bromophenyl-2-2-2-trifluoroethan-1-amine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S88021
Availability details for
Quantity
  • Availability
  • Add to Cart