ADVH49B8342B AChemBlock

SQ-609

  • Cas Number : 627052-25-9
  • Molecular Weight : 346.56

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/sq-609.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S74751
Availability details for
Quantity
  • Availability
  • Add to Cart