ADVH49B8342F AChemBlock

(S)-1-(2,4-dimethylphenyl)ethan-1-amine

  • Cas Number : 856563-12-7
  • Molecular Weight : 149.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/s-1-2-4-dimethylphenyl-ethan-1-amine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S88036
Availability details for
Quantity
  • Availability
  • Add to Cart