ADVH49B83437 AChemBlock

(R)-but-3-en-2-amine hydrochloride

  • Cas Number : 869485-70-1
  • Molecular Weight : 107.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/r-but-3-en-2-amine-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S88044
Availability details for
Quantity
  • Availability
  • Add to Cart