ADVH49B83438 AChemBlock

5-(trifluoromethyl)-2,3-dihydro-1H-inden-1-ol

  • Cas Number : 869725-57-5
  • Molecular Weight : 202.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/5-trifluoromethyl-2-3-dihydro-1h-inden-1-ol.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S88045
Availability details for
Quantity
  • Availability
  • Add to Cart