ADVH49B83439 AChemBlock

4-fluorothieno[2,3-c]pyridine-2-carboxylic acid

  • Cas Number : 870235-46-4
  • Molecular Weight : 197.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/4-fluorothieno-2-3-c-pyridine-2-carboxylic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S88046
Availability details for
Quantity
  • Availability
  • Add to Cart