ADVH49B8343C AChemBlock

2-amino-2-(2,4-difluorophenyl)ethan-1-ol

  • Cas Number : 872574-40-8
  • Molecular Weight : 173.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/2-amino-2-2-4-difluorophenyl-ethan-1-ol.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S88049
Availability details for
Quantity
  • Availability
  • Add to Cart