ADVH49B83444 AChemBlock

5-chloro-6-fluoro-2,3-dihydro-1H-inden-1-one

  • Cas Number : 881190-94-9
  • Molecular Weight : 184.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/5-chloro-6-fluoro-2-3-dihydro-1h-inden-1-one.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S88057
Availability details for
Quantity
  • Availability
  • Add to Cart