ADVH49B83446 AChemBlock

N-(4-methyl-3-nitrophenyl)-4-(trifluoromethyl)benzamide

  • Cas Number : 883028-23-7
  • Molecular Weight : 324.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/n-4-methyl-3-nitrophenyl-4-trifluoromethyl-benzamide.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S88059
Availability details for
Quantity
  • Availability
  • Add to Cart