ADVH49B83447 AChemBlock

1-(isoxazol-3-yl)ethan-1-one

  • Cas Number : 88511-37-9
  • Molecular Weight : 111.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/1-isoxazol-3-yl-ethan-1-one.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S88060
Availability details for
Quantity
  • Availability
  • Add to Cart