ADVH49B83449 AChemBlock

1-(3-chlorophenyl)-2,2,2-trifluoroethan-1-amine

  • Cas Number : 886368-66-7
  • Molecular Weight : 209.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/1-3-chlorophenyl-2-2-2-trifluoroethan-1-amine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S88062
Availability details for
Quantity
  • Availability
  • Add to Cart