ADVH49B8344A AChemBlock

3-(3-chloro-2-fluorophenyl)propanoic acid

  • Cas Number : 886498-12-0
  • Molecular Weight : 202.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/3-3-chloro-2-fluorophenyl-propanoic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S88063
Availability details for
Quantity
  • Availability
  • Add to Cart