ADVH49B8344D AChemBlock

(4-bromo-2,6-difluorophenyl)methanamine

  • Cas Number : 887585-99-1
  • Molecular Weight : 222.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/4-bromo-2-6-difluorophenyl-methanamine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S88066
Availability details for
Quantity
  • Availability
  • Add to Cart