ADVH49B8344E AChemBlock

7-fluoronaphthalen-2-ol

  • Cas Number : 889884-94-0
  • Molecular Weight : 162.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/7-fluoronaphthalen-2-ol.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S88067
Availability details for
Quantity
  • Availability
  • Add to Cart