ADVH49B8344F AChemBlock

ethyl 2-(2,4,6-trichloropyrimidin-5-yl)acetate

  • Cas Number : 889944-72-3
  • Molecular Weight : 269.51

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/ethyl-2-2-4-6-trichloropyrimidin-5-yl-acetate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S88068
Availability details for
Quantity
  • Availability
  • Add to Cart