ADVH49B83453 AChemBlock

8-oxo-5,6,7,8-tetrahydronaphthalene-2-carboxylic acid

  • Cas Number : 89781-52-2
  • Molecular Weight : 190.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/8-oxo-5-6-7-8-tetrahydronaphthalene-2-carboxylic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S88072
Availability details for
Quantity
  • Availability
  • Add to Cart