ADVH49B83454 AChemBlock

cyclopentyl(3,4-dichlorophenyl)methanone

  • Cas Number : 898791-87-2
  • Molecular Weight : 243.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/cyclopentyl-3-4-dichlorophenyl-methanone.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S88073
Availability details for
Quantity
  • Availability
  • Add to Cart