ADVH49B83458 AChemBlock

(S)-2-amino-3-(3,5-dimethylphenyl)propanoic acid hydrochloride

  • Cas Number : 906365-71-7
  • Molecular Weight : 229.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/s-2-amino-3-3-5-dimethylphenyl-propanoic-acid-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S88077
Availability details for
Quantity
  • Availability
  • Add to Cart