ADVH49B8345B AChemBlock

4,5-difluoro-2,3-dihydro-1H-inden-1-amine

  • Cas Number : 907973-34-6
  • Molecular Weight : 169.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/4-5-difluoro-2-3-dihydro-1h-inden-1-amine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S88080
Availability details for
Quantity
  • Availability
  • Add to Cart