ADVH49B8345E AChemBlock

(R)-4-(1-aminoethyl)benzonitrile hydrochloride

  • Cas Number : 911372-78-6
  • Molecular Weight : 182.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/r-4-1-aminoethyl-benzonitrile-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S88083
Availability details for
Quantity
  • Availability
  • Add to Cart