ADVH49B83461 AChemBlock

6-bromonaphthalen-1-ol

  • Cas Number : 91270-68-7
  • Molecular Weight : 223.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/6-bromonaphthalen-1-ol.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S88086
Availability details for
Quantity
  • Availability
  • Add to Cart