ADVH49B83462 AChemBlock

methyl 4-oxochromane-8-carboxylate

  • Cas Number : 91344-89-7
  • Molecular Weight : 206.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/methyl-4-oxochromane-8-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S88087
Availability details for
Quantity
  • Availability
  • Add to Cart