ADVH49B83463 AChemBlock

2-([1,1'-biphenyl]-3-yl)pyrrolidine

  • Cas Number : 914299-84-6
  • Molecular Weight : 223.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/2-1-1-biphenyl-3-yl-pyrrolidine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S88088
Availability details for
Quantity
  • Availability
  • Add to Cart