ADVH49B83464 AChemBlock

1-(4-(tert-butyl)phenyl)ethan-1-amine hydrochloride

  • Cas Number : 91552-71-5
  • Molecular Weight : 213.75

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/1-4-tert-butyl-phenyl-ethan-1-amine-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S88089
Availability details for
Quantity
  • Availability
  • Add to Cart