ADVH49B83468 AChemBlock

(S)-4-(1-aminoethyl)benzoic acid hydrochloride

  • Cas Number : 916211-64-8
  • Molecular Weight : 201.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/s-4-1-aminoethyl-benzoic-acid-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S88093
Availability details for
Quantity
  • Availability
  • Add to Cart