ADVH49B83469 AChemBlock

(S)-5,6-difluoro-2,3-dihydro-1H-inden-1-amine

  • Cas Number : 916214-29-4
  • Molecular Weight : 169.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/s-5-6-difluoro-2-3-dihydro-1h-inden-1-amine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S88094
Availability details for
Quantity
  • Availability
  • Add to Cart