ADVH49B83482 AChemBlock

tert-butyl (S)-(1-(3-fluorophenyl)-3-hydroxypropan-2-yl)carbamate

  • Cas Number : 944470-56-8
  • Molecular Weight : 269.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-s-1-3-fluorophenyl-3-hydroxypropan-2-yl-carbamate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S88119
Availability details for
Quantity
  • Availability
  • Add to Cart