ADVH49B83484 AChemBlock

(R)-2-amino-2-(3,5-difluorophenyl)ethan-1-ol

  • Cas Number : 957121-38-9
  • Molecular Weight : 173.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/r-2-amino-2-3-5-difluorophenyl-ethan-1-ol.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S88121
Availability details for
Quantity
  • Availability
  • Add to Cart