ADVH49B83487 AChemBlock

6-bromo-4,4-dimethyl-3,4-dihydronaphthalen-1(2H)-one

  • Cas Number : 98453-60-2
  • Molecular Weight : 253.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/6-bromo-4-4-dimethyl-3-4-dihydronaphthalen-1-2h-one.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S88124
Availability details for
Quantity
  • Availability
  • Add to Cart