ADVH49B83488 AChemBlock

benzo[b]thiophen-4-ol

  • Molecular Weight : 150.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/benzo-b-thiophen-4-ol.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S88125
Availability details for
Quantity
  • Availability
  • Add to Cart