ADVH49B8348A AChemBlock

morpholin-3-ylmethanol

  • Cas Number : 106910-83-2
  • Molecular Weight : 117.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/morpholin-3-ylmethanol.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S88140
Availability details for
Quantity
  • Availability
  • Add to Cart