ADVH49B8348C AChemBlock

3-(bromomethyl)-5-methylpyridine hydrobromide

  • Cas Number : 1235342-53-6
  • Molecular Weight : 266.96

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/3-bromomethyl-5-methylpyridine-hydrobromide.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S88164
Availability details for
Quantity
  • Availability
  • Add to Cart