ADVH49B834C4 AChemBlock

tert-butyl (R)-3-vinylmorpholine-4-carboxylate

  • Molecular Weight : 213.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-r-3-vinylmorpholine-4-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S88258
Availability details for
Quantity
  • Availability
  • Add to Cart