ADVH49B834C8 AChemBlock

dimethyl ((3-((tert-butyldimethylsilyl)oxy)phenyl)(methoxy)methyl)phosphonate

  • Molecular Weight : 360.46

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/dimethyl-3-tert-butyldimethylsilyl-oxy-phenyl-methoxy-methyl-phosphonate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S88646
Availability details for
Quantity
  • Availability
  • Add to Cart