ADVH49B834C9 AChemBlock

N,N,N-Tris(4-aminobutyl)amine Tetrahydrochloride

  • Molecular Weight : 376.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/n-n-n-tris-4-aminobutyl-amine-tetrahydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S88647
Availability details for
Quantity
  • Availability
  • Add to Cart