ADVH49B834CA AChemBlock

(3-((Z)-((1r,3s,5R,7S)-adamantan-2-ylidene)(methoxy)methyl)-2-chlorophenoxy)(tert-butyl)dimethylsilane

  • Molecular Weight : 419.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/3-z-1r-3s-5r-7s-adamantan-2-ylidene-methoxy-methyl-2-chlorophenoxy-tert-butyl-dimethylsilane.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S88648
Availability details for
Quantity
  • Availability
  • Add to Cart