ADVH49B834CE AChemBlock

perfluorophenyl 2-(4-methoxyphenyl)acetate

  • Molecular Weight : 332.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/perfluorophenyl-2-4-methoxyphenyl-acetate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S88658
Availability details for
Quantity
  • Availability
  • Add to Cart