ADVH49B834D1 AChemBlock

methyl ((1-tosyl-1H-indol-3-yl)methyl) carbonate

  • Molecular Weight : 359.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/methyl-1-tosyl-1h-indol-3-yl-methyl-carbonate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S88663
Availability details for
Quantity
  • Availability
  • Add to Cart