ADVH49B834D6 AChemBlock

N-(2-cyclopropylideneethyl)-4-methyl-N-(2-(1-methyl-1H-indol-3-yl)ethyl)benzenesulfonamide

  • Molecular Weight : 394.53

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/n-2-cyclopropylideneethyl-4-methyl-n-2-1-methyl-1h-indol-3-yl-ethyl-benzenesulfonamide.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S88674
Availability details for
Quantity
  • Availability
  • Add to Cart