ADVH49B834D8 AChemBlock

ethyl 3,6-bis(2,3-dichlorophenyl)-5-methylpyrazine-2-carboxylate

  • Molecular Weight : 456.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/ethyl-3-6-bis-2-3-dichlorophenyl-5-methylpyrazine-2-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S88688
Availability details for
Quantity
  • Availability
  • Add to Cart