ADVH49B834DA AChemBlock

(S)-6,6'-di(naphthalen-2-yl)-2,2',3,3'-tetrahydro-1,1'-spirobi[indene]-7,7'-diol

  • Molecular Weight : 504.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/s-6-6-di-naphthalen-2-yl-2-2-3-3-tetrahydro-1-1-spirobi-indene-7-7-diol.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S88690
Availability details for
Quantity
  • Availability
  • Add to Cart