ADVH49B834DB AChemBlock

tert-butyl 2,4-difluoro-3-hydroxy-8-azabicyclo[3.2.1]octane-8-carboxylate

  • Molecular Weight : 263.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-2-4-difluoro-3-hydroxy-8-azabicyclo-3-2-1-octane-8-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S88842
Availability details for
Quantity
  • Availability
  • Add to Cart